7-(Trifluoromethyl)benzothiophene-2-carboxamide
Supplier: Apollo Scientific
Warning
PC57780-1G PC57780-250MG
APOSPC57780-1GEA 132 EUR
APOSPC57780-1G APOSPC57780-250MG
7-(Trifluoromethyl)benzothiophene-2-carboxamide
7-(Trifluoromethyl)benzothiophene-2-carboxamide
Formula: C₁₀H₆F₃NOS
MW: 245.221 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD29922223
CAS Number: 1956369-80-4