7-(Trifluoromethyl)benzothiophene-2-carbonitrile
Supplier: Apollo Scientific
Warning
PC57781-1G PC57781-250MG
APOSPC57781-1GEA 136 EUR
APOSPC57781-1G APOSPC57781-250MG
7-(Trifluoromethyl)benzothiophene-2-carbonitrile
7-(Trifluoromethyl)benzothiophene-2-carbonitrile
Formula: C₁₀H₄F₃NS
MW: 227.206 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD31567883
CAS Number: 2115921-53-2