2-(Bromomethyl)-5-(trifluoromethyl)benzothiophene
Supplier: Apollo Scientific
Danger
PC57789-1G PC57789-250MG
APOSPC57789-1GEA 136 EUR
APOSPC57789-1G APOSPC57789-250MG
2-(Bromomethyl)-5-(trifluoromethyl)benzothiophene
2-(Bromomethyl)-5-(trifluoromethyl)benzothiophene
Formula: C₁₀H₆BrF₃S
MW: 295.119 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD28399005
CAS Number: 1858256-40-2
UN: 3261
ADR: 8,III