N-(Pentafluoropropionyl)imidazole 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2,3,3,3-Pentafluoro-1-(1H-imidazol-1-yl)propan-1-one
PC5784-25G PC5784-5G PC5784-1G PC5784-10G
APOSPC5784-25GEA 619 EUR
APOSPC5784-25G APOSPC5784-5G APOSPC5784-1G APOSPC5784-10G
N-(Pentafluoropropionyl)imidazole 98%
N-(Pentafluoropropionyl)imidazole
Formula: C₆H₃F₅N₂O
MW: 214.093 g/mol
Boiling Pt: 46…47 °C (15 mm Hg)
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00014502
CAS Number: 71735-32-5
EINECS: 275-944-1
UN: 3261
ADR: 8,III