2-Bromo-3-(trifluoromethoxy)benzoic acid
Supplier: Apollo Scientific
Warning
PC5797-1G PC5797-250MG
APOSPC5797-1GEA 348 EUR
APOSPC5797-1G APOSPC5797-250MG
2-Bromo-3-(trifluoromethoxy)benzoic acid
2-Bromo-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD12026457
CAS Number: 1159512-46-5