1-[2-(Trifluoromethyl)quinol-4-yl]piperazine 97%
Supplier: Apollo Scientific
Warning
PC5892-1G PC5892-250MG
APOSPC5892-1GEA 203 EUR
APOSPC5892-1G APOSPC5892-250MG
1-[2-(Trifluoromethyl)quinol-4-yl]piperazine 97%
1-[2-(Trifluoromethyl)quinol-4-yl]piperazine
Formula: C₁₄H₁₄F₃N₃
MW: 281.276 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00219765
CAS Number: 175203-79-9