2-Fluoro-5-(trifluoromethoxy)benzoyl chloride 98%
Supplier: Apollo Scientific
Danger
PC5897-25G PC5897-5G PC5897-1G
APOSPC5897-25GEA 540 EUR
APOSPC5897-25G APOSPC5897-5G APOSPC5897-1G
2-Fluoro-5-(trifluoromethoxy)benzoyl chloride 98%
2-Fluoro-5-(trifluoromethoxy)benzoyl chloride
Formula: C₈H₃ClF₄O₂
MW: 242.555 g/mol
Boiling Pt: 191 °C
Density: 1.499 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD04115888
CAS Number: 886497-89-8
UN: 3265
ADR: 8,II