3-Nonafluorobutyl-2-iodopropanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Iodo-1H,1H,2H,3H,3H-nonafluoroheptan-1-ol
PC5906GB-5G PC5906GB-1G PC5906GB-250MG
APOSPC5906GB-5GEA 301 EUR
APOSPC5906GB-5G APOSPC5906GB-1G APOSPC5906GB-250MG
3-Nonafluorobutyl-2-iodopropanol 97%
3-Nonafluorobutyl-2-iodopropanol
Formula: C₇H₆F₉IO
MW: 404.012 g/mol
Boiling Pt: 56…58 °C (1 mm Hg)
Density: 1.972 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236068
CAS Number: 80233-96-1