(1S)-N-Methyl-1-[2-(trifluoromethyl)phenyl]ethylamine
Supplier: Apollo Scientific
Synonyms: (1S)-N-{1-[2-(Trifluoromethyl)phenyl]ethyl}methylamine
PC5962-1G PC5962-250MG
APOSPC5962-1GEA 654 EUR
APOSPC5962-1G APOSPC5962-250MG
(1S)-N-Methyl-1-[2-(trifluoromethyl)phenyl]ethylamine
(1S)-N-Methyl-1-[2-(trifluoromethyl)phenyl]ethylamine
Formula: C₁₀H₁₂F₃N
MW: 203.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779102
CAS Number: 1212332-00-7