1H,1H,2H,2H-Henicosafluorododec-1-yl methacrylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Henicosafluorododec-1-yl 2-methylprop-2-enoate, 2-(Perfluorodec-1-yl)ethyl methacrylate
PC5969P-1G PC5969P-5G
APOSPC5969P-1GEA 58 EUR
APOSPC5969P-1G APOSPC5969P-5G
1H,1H,2H,2H-Henicosafluorododec-1-yl methacrylate 97%
1H,1H,2H,2H-Henicosafluorododec-1-yl methacrylate
Formula: C₁₆H₉F₂₁O₂
MW: 632.208 g/mol
Boiling Pt: 95…100 °C (0.1 mm Hg)
Melting Pt: 40…45 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236095
CAS Number: 2144-54-9
EINECS: 218-408-4