2-Phenyl-2-(trifluoroacetyl)acetonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Oxo-2-pheny-4,4,4-trifluorolbutanenitrile, α-(Trifluoroacetyl)benzeneacetonitrile
PC6223G-1G PC6223G-5G
APOSPC6223G-1GEA 40.6 EUR
APOSPC6223G-1G APOSPC6223G-5G
2-Phenyl-2-(trifluoroacetyl)acetonitrile 97%
2-Phenyl-2-(trifluoroacetyl)acetonitrile
Formula: C₁₀H₆F₃NO
MW: 213.156 g/mol
Melting Pt: 85…87 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00077582
CAS Number: 492-16-0
UN: 3439
ADR: 6.1,III