3-Fluoro-4-(methylsulphonyl)aniline
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-2-fluorophenyl methyl sulphone
PC6249-1G PC6249-5G PC6249-10G
APOSPC6249-1GEA 49.3 EUR
APOSPC6249-1G APOSPC6249-5G APOSPC6249-10G
3-Fluoro-4-(methylsulphonyl)aniline
3-Fluoro-4-(methylsulphonyl)aniline
Formula: C₇H₈FNO₂S
MW: 189.207 g/mol
Melting Pt: 117…118.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264555
CAS Number: 252561-34-5