2-Fluoro-3-(trifluoromethoxy)benzoic acid
Supplier: Apollo Scientific
Warning
PC6353-1G PC6353-5G PC6353-25G
APOSPC6353-1GEA 49.3 EUR
APOSPC6353-1G APOSPC6353-5G APOSPC6353-25G
2-Fluoro-3-(trifluoromethoxy)benzoic acid
2-Fluoro-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
MW: 224.109 g/mol
Melting Pt: 124…125 °C
Storage Temperature: Ambient
MDL Number: MFCD12026477
CAS Number: 1159512-62-5