4-Bromo-2-chloro-5-fluorobenzoyl chloride 95%
Supplier: Apollo Scientific
Danger
PC6589-1G PC6589-250MG PC6589-5G
APOSPC6589-1GEA 209 EUR
APOSPC6589-1G APOSPC6589-250MG APOSPC6589-5G
4-Bromo-2-chloro-5-fluorobenzoyl chloride 95%
4-Bromo-2-chloro-5-fluorobenzoyl chloride
Formula: C₇H₂BrCl₂FO
MW: 271.899 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236590
CAS Number: 203302-93-6
UN: 3265
ADR: 8,II