(2R)-2-Fluoropropan-1-ol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (2R)-2-Fluoro-1-hydroxypropane
PC6608-1G PC6608-250MG
APOSPC6608-1GEA 1250 EUR
APOSPC6608-1G APOSPC6608-250MG
(2R)-2-Fluoropropan-1-ol 97%
(2R)-2-Fluoropropan-1-ol
Formula: C₃H₇FO
MW: 78.085 g/mol
Boiling Pt: 115 °C
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD09763597
CAS Number: 876747-18-1
UN: 1987
ADR: 3,III