5,5,5-Trifluoro-4-(trifluoromethyl)pent-3-en-2-one 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Oxo-1,1,1-trifluoro-2-(trifluoromethyl)pent-2-ene, 4,4-Bis(trifluoromethyl)but-3-en-2-one
PC6653-1G PC6653-5G
APOSPC6653-1GEA 114 EUR
APOSPC6653-1G APOSPC6653-5G
5,5,5-Trifluoro-4-(trifluoromethyl)pent-3-en-2-one 97+%
5,5,5-Trifluoro-4-(trifluoromethyl)pent-3-en-2-one
Formula: C₆H₄F₆O
MW: 206.086 g/mol
Boiling Pt: 68 °C (135 mm Hg)
Density: 1.4102 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09763630
CAS Number: 1422-36-2
UN: 1993
ADR: 3,III