4-(2,2,2-Trifluoroethoxy)nitrobenzene 97%
Supplier: Apollo Scientific
Warning
PC6675-1G PC6675-5G
APOSPC6675-1GEA 34.8 EUR
APOSPC6675-1G APOSPC6675-5G
4-(2,2,2-Trifluoroethoxy)nitrobenzene 97%
4-(2,2,2-Trifluoroethoxy)nitrobenzene
Formula: C₈H₆F₃NO₃
MW: 221.133 g/mol
Boiling Pt: 70…73 °C (3 mm Hg)
Melting Pt: 76…78 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00077608
CAS Number: 62149-35-3