Perfluoro(4,4'-diphenoxybiphenyl) 90%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4'-Bis(pentafluorophenoxy)octafluorobiphenyl
PC6682-1G PC6682-250MG
APOSPC6682-1GEA 466 EUR
APOSPC6682-1G APOSPC6682-250MG
Perfluoro(4,4'-diphenoxybiphenyl) 90%
Perfluoro(4,4'-diphenoxybiphenyl)
Formula: C₂₄F₁₈O₂
MW: 662.227 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09763620
CAS Number: 14055-56-2