2',3',4',5'-Tetrafluoroacetophenone 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,3,4,5-Tetrafluorophenyl)ethan-1-one, 1-(2,3,4,5-Tetrafluorophenyl)ethanone, 1-Oxo-1-(2,3,4,5-tetrafluorophenyl)ethane
PC6693-1G PC6693-5G
APOSPC6693-1GEA 35 EUR
APOSPC6693-1G APOSPC6693-5G
2',3',4',5'-Tetrafluoroacetophenone 99%
2',3',4',5'-Tetrafluoroacetophenone
Formula: C₈H₄F₄O
Boiling Pt: 83…84 °C (20 mm Hg)
Density: 1.408 g/cm³ (25 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00134310
CAS Number: 66286-21-3
UN: 2810
ADR: 6.1,III