Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate 95+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-amino-4-phenyl-5-(trifluoromethyl)-2-thenoate, 3-Amino-2-(methoxycarbonyl)-4-phenyl-5-(trifluoromethyl)thiophene
PC6719-1G PC6719-5G
APOSPC6719-1GEA 123 EUR
APOSPC6719-1G APOSPC6719-5G
Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate 95+%
Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Formula: C₁₃H₁₀F₃NO₂S
MW: 301.284 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00832742
CAS Number: 256353-38-5