4H-Tetrafluorophenyl methacrylate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3,5,6-Tetrafluorophenyl methacrylate, 2,3,5,6-Tetrafluorophenyl 2-methylprop-2-enoate
PC6744-25G
PC6744-5G
APOSPC6744-25GEA
379
EUR
APOSPC6744-25G
APOSPC6744-5G
4H-Tetrafluorophenyl methacrylate 97%
4H-Tetrafluorophenyl methacrylate
Formula:
C₁₀H₆F₄O₂ MW: 234.147 g/mol Density: 1.3 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00129959 CAS Number: 101156-31-4 |