Phenyl trifluorovinyl ether
Supplier: Apollo Scientific
Warning
Synonyms: [(Trifluorovinyl)oxy]benzene
PC6751-1G PC6751-5G
APOSPC6751-1GEA 286 EUR
APOSPC6751-1G APOSPC6751-5G
Phenyl trifluorovinyl ether
Phenyl trifluorovinyl ether
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 134…134.5 °C
Melting Pt: –23.5…–22 °C
Density: 1.25 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10567162
CAS Number: 772-53-2
UN: 1993
ADR: 3,III