4,5-Difluoro-2-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Warning
PC6789-250MG PC6789-50MG
APOSPC6789-250MGEA 418 EUR
APOSPC6789-250MG APOSPC6789-50MG
4,5-Difluoro-2-(trifluoromethyl)-1H-benzimidazole
4,5-Difluoro-2-(trifluoromethyl)-1H-benzimidazole
Formula: C₈H₃F₅N₂
Storage Temperature: Ambient
MDL Number: MFCD00665872
CAS Number: 28196-83-0