3-Iodo-1,2,4,5-tetrafluorobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Iodo-4H-tetrafluorobenzene, 1,2,4,5-tetrafluoro-3-iodobenzene, 1,2,4,5-Tetrafluoro-3-iodobenzene, 4-Iodo-1H-tetrafluorobenzene
PC6815-1G PC6815-250MG
APOSPC6815-1GEA 546 EUR
APOSPC6815-1G APOSPC6815-250MG
3-Iodo-1,2,4,5-tetrafluorobenzene 97%
3-Iodo-1,2,4,5-tetrafluorobenzene
Formula: C₆HF₄I
MW: 275.97 g/mol
Density: 2.1 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09763612
CAS Number: 5243-24-3