1,3-Bis(1H,1H,5H-perfluoropentoxy)propan-2-ol
Supplier: Apollo Scientific
Synonyms: 1,3-Bis[(2,2,3,3,4,4,5,5-octafluoropentyl)oxy]propan-2-ol
PC6899-25G PC6899-5G
APOSPC6899-25GEA 344 EUR
APOSPC6899-25G APOSPC6899-5G
1,3-Bis(1H,1H,5H-perfluoropentoxy)propan-2-ol
1,3-Bis(1H,1H,5H-perfluoropentoxy)propan-2-ol
Formula: C₁₃H₁₂F₁₆O₃
MW: 520.207 g/mol
Boiling Pt: 178 °C (0.8 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00337849
CAS Number: 929080-18-2