2,3-Difluoro-4-(trifluoromethyl)benzaldehyde 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Difluoro-4-formylbenzotrifluoride
PC6951-1G PC6951-5G PC6951-25G
APOSPC6951-1GEA 31.9 EUR
APOSPC6951-1G APOSPC6951-5G APOSPC6951-25G
2,3-Difluoro-4-(trifluoromethyl)benzaldehyde 95+%
2,3-Difluoro-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₃F₅O
MW: 210.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236254
CAS Number: 134099-20-0