1,2-Dihydro-2-oxo-6-(trifluoromethyl)pyridine-3-carbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Oxo-6-(trifluoromethyl)-1,2-dihydropyridine-3-carbonitrile, 3-Cyano-1,2-dihydro-2-oxo-6-(trifluoromethyl)pyridine
PC6995-1G PC6995-250MG
APOSPC6995-1GEA 517 EUR
APOSPC6995-1G APOSPC6995-250MG
1,2-Dihydro-2-oxo-6-(trifluoromethyl)pyridine-3-carbonitrile
1,2-Dihydro-2-oxo-6-(trifluoromethyl)pyridine-3-carbonitrile
Formula: C₇H₃F₃N₂O
MW: 188.107 g/mol
Melting Pt: 210…211 °C
Storage Temperature: Ambient
MDL Number: MFCD09749653
CAS Number: 116548-04-0