4-Hydroxy-2-methyl-8-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
Synonyms: 2-Methyl-8-(trifluoromethyl)quinolin-4-ol
PC7148-1G PC7148-250MG
APOSPC7148-1GEA 201 EUR
APOSPC7148-1G APOSPC7148-250MG
4-Hydroxy-2-methyl-8-(trifluoromethyl)quinoline
4-Hydroxy-2-methyl-8-(trifluoromethyl)quinoline
Formula: C₁₁H₈F₃NO
MW: 227.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100260
CAS Number: 140908-88-9