2,3-Difluorophenyl trifluoromethanesulphonate 95%
Supplier: Apollo Scientific
PC7162-1G PC7162-5G
APOSPC7162-1GEA 141 EUR
APOSPC7162-1G APOSPC7162-5G
2,3-Difluorophenyl trifluoromethanesulphonate 95%
2,3-Difluorophenyl trifluoromethanesulphonate
Formula: C₇H₃F₅O₃S
MW: 262.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784320
CAS Number: 211315-75-2