2-Amino-7-fluoro-3-methylquinoline
Supplier: Apollo Scientific
Warning
Synonyms: 7-Fluoro-3-methylquinolin-2-amine, 2-Amino-7-fluoro-3-methyl-1-azanaphthalene
PC7195-1G PC7195-250MG
APOSPC7195-1GEA 582 EUR
APOSPC7195-1G APOSPC7195-250MG
2-Amino-7-fluoro-3-methylquinoline
2-Amino-7-fluoro-3-methylquinoline
Formula: C₁₀H₉FN₂
MW: 176.19 g/mol
Melting Pt: 203…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00272513
CAS Number: 203506-29-0