Methyl 3-(cyclopropylamino)-2-(2,4-dichloro-5-fluorobenzoyl)acrylate
Supplier: Apollo Scientific
Warning
Synonyms: {[2-(2,4-Dichloro-5-fluorobenzoyl)-3-methoxy-3-oxoprop-1-en-1-yl]amino}cyclopropane, Methyl 3-(cyclopropylamino)-2-(2,4-dichloro-5-fluorobenzoyl)prop-2-enoate
PC7235-100G PC7235-25G PC7235-5G
APOSPC7235-100GEA 418 EUR
APOSPC7235-100G APOSPC7235-25G APOSPC7235-5G
Methyl 3-(cyclopropylamino)-2-(2,4-dichloro-5-fluorobenzoyl)acrylate
Methyl 3-(cyclopropylamino)-2-(2,4-dichloro-5-fluorobenzoyl)acrylate
Formula: C₁₄H₁₂Cl₂FNO₃
MW: 332.154 g/mol
Melting Pt: 150…154 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02683487
CAS Number: 105392-26-5