1,1,1,2,2-Pentafluoro-3-methylbutan-3-ol
Supplier: Apollo Scientific
Warning
PC7242-1G PC7242-250MG PC7242-5G
APOSPC7242-1GEA 358 EUR
APOSPC7242-1G APOSPC7242-250MG APOSPC7242-5G
1,1,1,2,2-Pentafluoro-3-methylbutan-3-ol
1,1,1,2,2-Pentafluoro-3-methylbutan-3-ol
Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 90…92 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00069121
CAS Number: 374-46-9
UN: 1993
ADR: 3,II