4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-5-bromo-trifluoromethoxybenzene, 5-Bromo-2-fluorophenyl trifluoromethyl ether, 5-Bromo-α,α,α,2-tetrafluoroanisole
PC7291-1G PC7291-5G PC7291-250MG PC7291-25G
APOSPC7291-1GEA 56.7 EUR
APOSPC7291-1G APOSPC7291-5G APOSPC7291-250MG APOSPC7291-25G
4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene 99%
4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₄O
MW: 258.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660160
CAS Number: 886496-45-3
UN: 1993
ADR: 3 III