2-Deoxy-2-fluoro-D-mannose
Supplier: Apollo Scientific
Warning
PC7309-100MG PC7309-10MG PC7309-25MG
APOSPC7309-100MGEA 790 EUR
APOSPC7309-100MG APOSPC7309-10MG APOSPC7309-25MG
2-Deoxy-2-fluoro-D-mannose
2-Deoxy-2-fluoro-D-mannose
Formula: C₆H₁₁FO₅
Melting Pt: 123…128 °C
Storage Temperature: Freezer
MDL Number: MFCD00797602
CAS Number: 31077-88-0