1,2-Bis[3-(trifluoromethyl)phenyl]-1,1,2,2-tetrafluoroethane 97%
Supplier: Apollo Scientific
Synonyms: 1,1'-(1,1,2,2-Tetrafluoroethane-1,2-diyl)bis[3-(trifluoromethyl)benzene]
PC7397-1G PC7397-25G PC7397-5G
APOSPC7397-1GEA 102 EUR
APOSPC7397-1G APOSPC7397-25G APOSPC7397-5G
1,2-Bis[3-(trifluoromethyl)phenyl]-1,1,2,2-tetrafluoroethane 97%
1,2-Bis[3-(trifluoromethyl)phenyl]-1,1,2,2-tetrafluoroethane
Formula: C₁₆H₈F₁₀
MW: 390.219 g/mol
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD01631640
CAS Number: 287172-67-2