4-(Trifluoromethoxy)phenoxyacetic hydrazide 97%
Supplier: Apollo Scientific
|
Warning
|
PC7439PC-1G
PC7439PC-250MG
APOSPC7439PC-1GEA
69.6
EUR
APOSPC7439PC-1G
APOSPC7439PC-250MG
4-(Trifluoromethoxy)phenoxyacetic hydrazide 97%
4-(Trifluoromethoxy)phenoxyacetic hydrazide
Formula:
C₉H₉F₃N₂O₃ Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052355 CAS Number: 175204-36-1 |