(1S)-1-(4-Fluorophenyl)-N-methylethylamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: (1S)-1-(4-Fluorophenyl)-N-methylethanamine
PC7545-100MG PC7545-1G PC7545-250MG
APOSPC7545-100MGEA 115 EUR
APOSPC7545-100MG APOSPC7545-1G APOSPC7545-250MG
(1S)-1-(4-Fluorophenyl)-N-methylethylamine 98%
(1S)-1-(4-Fluorophenyl)-N-methylethylamine
Formula: C₉H₁₂FN
MW: 153.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779100
CAS Number: 672906-67-1