3,5-Difluoro-2-nitroanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1,5-Difluoro-3-methoxy-2-nitrobenzene, 3,5-Difluoro-2-nitrophenyl methyl ether
PC7672-1G PC7672-250MG PC7672-5G
APOSPC7672-1GEA 102 EUR
APOSPC7672-1G APOSPC7672-250MG APOSPC7672-5G
3,5-Difluoro-2-nitroanisole
3,5-Difluoro-2-nitroanisole
Formula: C₇H₅F₂NO₃
MW: 189.116 g/mol
Melting Pt: 59.5…59.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11865203
CAS Number: 66684-61-5