4-Bromo-1,1,1-trifluoro-2-(trifluoromethyl)butane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,4-Trifluoro-3-(trifluoromethyl)butyl bromide, 1,1-Bis(trifluoromethyl)-3-bromopropane
PC7854H-1G PC7854H-5G
APOSPC7854H-1GEA 200 EUR
APOSPC7854H-1G APOSPC7854H-5G
4-Bromo-1,1,1-trifluoro-2-(trifluoromethyl)butane 97%
4-Bromo-1,1,1-trifluoro-2-(trifluoromethyl)butane
Formula: C₅H₅BrF₆
MW: 258.988 g/mol
Boiling Pt: 108 °C
Density: 1.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236629
CAS Number: 203303-02-0