1H,1H,2H,2H-Perfluorotetradecyltrichlorosilane 97%
Supplier: Apollo Scientific
Danger
Synonyms: Trichloro-1H,1H,2H,2H-perfluorotetradecylsilane
PC7902-1G PC7902-5G
APOSPC7902-1GEA 0 EUR
APOSPC7902-1G APOSPC7902-5G
1H,1H,2H,2H-Perfluorotetradecyltrichlorosilane 97%
1H,1H,2H,2H-Perfluorotetradecyltrichlorosilane
Formula: C₁₄H₄Cl₃F₂₅Si
MW: 781.586 g/mol
Boiling Pt: 114…115 °C (1.2 mm Hg)
Melting Pt: 91…93 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD07368764
CAS Number: 102488-50-6
UN: 1759
ADR: 8 II