(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine
Supplier: Apollo Scientific
Warning
PC8013-1G PC8013-250MG
APOSPC8013-1GEA 331 EUR
APOSPC8013-1G APOSPC8013-250MG
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine
Formula: C₁₀H₉F₆N
MW: 257.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093012
CAS Number: 127733-47-5