2-(Trifluoromethyl)anisole
Supplier: Apollo Scientific
Warning
PC8230-1G PC8230-250MG PC8230-5G
APOSPC8230-1GEA 87 EUR
APOSPC8230-1G APOSPC8230-250MG APOSPC8230-5G
2-(Trifluoromethyl)anisole
2-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Melting Pt: –14.3…–14.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00142938
CAS Number: 395-48-2
UN: 1993
ADR: 3 III