4-Bromo-2,6-difluorobenzyl bromide 98%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Bromo-2-(bromomethyl)-1,3-difluorobenzene, α,4-Dibromo-2,6-difluorotoluene
PC8241-1G PC8241-5G PC8241-25G
APOSPC8241-1GEA 84.1 EUR
APOSPC8241-1G APOSPC8241-5G APOSPC8241-25G
4-Bromo-2,6-difluorobenzyl bromide 98%
4-Bromo-2,6-difluorobenzyl bromide
Formula: C₇H₄Br₂F₂
MW: 285.911 g/mol
Boiling Pt: 243 °C
Density: 1.991 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094462
CAS Number: 162744-60-7
UN: 3261
ADR: 8,II