4-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine, N1-BOC protected 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(tert-Butoxycarbonyl)-4-[2-nitro-4-(trifluoromethyl)phenyl]homopiperazine, tert-Butyl 4-[2-nitro-4-(trifluoromethyl)phenyl]-1,4-diazepane-1-carboxylate
PC8327-1G PC8327-5G PC8327-250MG
APOSPC8327-1GEA 176 EUR
APOSPC8327-1G APOSPC8327-5G APOSPC8327-250MG
4-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine, N1-BOC protected 97%
4-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine, N1-BOC protected
Formula: C₁₇H₂₂F₃N₃O₄
MW: 389.369 g/mol
Melting Pt: 72…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00729028
CAS Number: 306934-72-5