2-(4-Fluorobenzylthio)ethylamine 95%
Supplier: Apollo Scientific
Warning
PC8344-100MG PC8344-500MG
APOSPC8344-100MGEA 221 EUR
APOSPC8344-100MG APOSPC8344-500MG
2-(4-Fluorobenzylthio)ethylamine 95%
2-(4-Fluorobenzylthio)ethylamine
Formula: C₉H₁₂FNS
MW: 185.262 g/mol
Boiling Pt: 98 °C (0.5 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01764909
CAS Number: 143627-49-0