2-{1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-yl}acetohydrazide 97%
Supplier: Apollo Scientific
PC8427-1G PC8427-5G
APOSPC8427-1GEA 212 EUR
APOSPC8427-1G APOSPC8427-5G
2-{1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-yl}acetohydrazide 97%
2-{1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-yl}acetohydrazide
Formula: C₁₃H₁₆ClF₃N₄O
MW: 336.741 g/mol
Melting Pt: 198…201 °C
Storage Temperature: Ambient
MDL Number: MFCD04038248
CAS Number: 537050-18-3