2-(2,4-Difluorophenoxy)-1-methyl-1H-indole-3-carboxaldehyde 97%
Supplier: Apollo Scientific
PC8455-1G PC8455-500MG
APOSPC8455-1GEA 533 EUR
APOSPC8455-1G APOSPC8455-500MG
2-(2,4-Difluorophenoxy)-1-methyl-1H-indole-3-carboxaldehyde 97%
2-(2,4-Difluorophenoxy)-1-methyl-1H-indole-3-carboxaldehyde
Formula: C₁₆H₁₁F₂NO₂
MW: 287.261 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD01568349
CAS Number: 773872-28-9