Ethyl 3-amino-3-[4-(trifluoromethoxy)anilino]prop-2-en-1-oate hydrochloride 97%
Supplier: Apollo Scientific
PC8465-1G PC8465-500MG
APOSPC8465-1GEA 977 EUR
APOSPC8465-1G APOSPC8465-500MG
Ethyl 3-amino-3-[4-(trifluoromethoxy)anilino]prop-2-en-1-oate hydrochloride 97%
Ethyl 3-amino-3-[4-(trifluoromethoxy)anilino]prop-2-en-1-oate hydrochloride
Formula: C₁₂H₁₃F₃N₂O₃·ClH
MW: 326.699 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00243911
CAS Number: 317821-98-0