Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate 97%
Supplier: Apollo Scientific
PC8533-1G PC8533-250MG
APOSPC8533-1GEA 220 EUR
APOSPC8533-1G APOSPC8533-250MG
Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate 97%
Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate
Formula: C₁₈H₁₃F₅O₄
MW: 388.285 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD09702368
CAS Number: 930110-97-7