Pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate 97%
Supplier: Apollo Scientific
PC8542-1G PC8542-250MG
APOSPC8542-1GEA 199 EUR
APOSPC8542-1G APOSPC8542-250MG
Pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate 97%
Pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate
Formula: C₁₄H₉F₅N₂O₃S
Melting Pt: 118…119.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09817538
CAS Number: 921939-02-8